Product Details
- SNP ID
-
rs2274253
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
64
- Location
-
Chr.1:117060534 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GCTTCTACGTGTGCCGGGCAGACAC[G/T]TGCAGCTTCGTGCGGGCCACCGAGT
- Phenotype
-
MIM: 604718
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929099
PubMed Links
Gene Details
- Gene
- LOC101929099
- Gene Name
- uncharacterized LOC101929099
There are no transcripts associated with this gene.
- Gene
- TTF2
- Gene Name
- transcription termination factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003594.3 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
NP_003585.3 |
XM_005271277.3 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_005271334.1 |
XM_011542303.2 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_011540605.1 |
XM_017002549.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858038.1 |
XM_017002550.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858039.1 |
XM_017002551.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858040.1 |
XM_017002552.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858041.1 |
XM_017002553.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858042.1 |
XM_017002554.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858043.1 |
XM_017002555.1 |
167 |
Silent Mutation |
ACG,ACT |
T36T |
XP_016858044.1 |
View Full Product Details