Product Details

SNP ID
rs2274330
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:8949411 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TGACAGGCTATGAGACCCAGGCAGG[C/G]GAGTTCCCCATGGTCAACAATGGCC
Phenotype
MIM: 114780
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CA6 PubMed Links

Gene Details

Gene
CA6
Gene Name
carbonic anhydrase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001215.3 329 Silent Mutation GGC,GGG G76G NP_001206.2
NM_001270500.1 329 Silent Mutation GGC,GGG G76G NP_001257429.1
NM_001270501.1 329 Intron NP_001257430.1
NM_001270502.1 329 Intron NP_001257431.1
XM_011542083.2 329 Silent Mutation GGC,GGG G80G XP_011540385.1
XM_011542084.2 329 Silent Mutation GGC,GGG G80G XP_011540386.1

View Full Product Details