Product Details

SNP ID
rs2274527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74727978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCAGATTTCATCCCATTTAAGTC[A/G]GTGGAACTTTTCTTTTGGAGTGACT
Phenotype
MIM: 607009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPM6 PubMed Links

Gene Details

Gene
TRPM6
Gene Name
transient receptor potential cation channel subfamily M member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177310.1 Intron NP_001170781.1
NM_001177311.1 Intron NP_001170782.1
NM_017662.4 Intron NP_060132.3
XM_011518244.1 Intron XP_011516546.1
XM_011518245.1 Intron XP_011516547.1
XM_011518246.1 Intron XP_011516548.1
XM_011518247.1 Intron XP_011516549.1
XM_011518248.1 Intron XP_011516550.1
XM_011518249.1 Intron XP_011516551.1
XM_011518250.1 Intron XP_011516552.1
XM_011518251.2 Intron XP_011516553.1
XM_011518252.2 Intron XP_011516554.1
XM_011518255.2 Intron XP_011516557.1
XM_017014287.1 Intron XP_016869776.1
XM_017014288.1 Intron XP_016869777.1
XM_017014289.1 Intron XP_016869778.1

View Full Product Details