Product Details

SNP ID
rs2274580
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:108561200 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCAGATGGGGAGCCCAGGCCCTGCC[C/T]TCATTTTAAAATGAGCCCCTGGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM102B PubMed Links

Gene Details

Gene
FAM102B
Gene Name
family with sequence similarity 102 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010883.2 Intron NP_001010883.2

View Full Product Details