Product Details

SNP ID
rs2287632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206738497 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTGATCTGTTAGGATTCCACGG[C/T]TTTGCCTTCAAATTCTGTAAAAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDH1B PubMed Links

Gene Details

Gene
MDH1B
Gene Name
malate dehydrogenase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039845.2 1807 Missense Mutation ACC,GCC T515A NP_001034934.1
NM_001282940.1 1807 Missense Mutation ACC,GCC T514A NP_001269869.1
XM_005246315.4 1807 Missense Mutation ACC,GCC T417A XP_005246372.1
XM_005246316.4 1807 Missense Mutation ACC,GCC T302A XP_005246373.1
XM_011510628.2 1807 Missense Mutation ACC,GCC T357A XP_011508930.1
XM_017003373.1 1807 Missense Mutation ACC,GCC T416A XP_016858862.1
XM_017003374.1 1807 Missense Mutation ACC,GCC T356A XP_016858863.1

View Full Product Details