Product Details

SNP ID
rs2977523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:133199167 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTGAGTGGCAAGCGGCAGCTTAC[A/G]GGCTTGGTTCTGGAGCAAGGACAGA
Phenotype
MIM: 603398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WISP1 PubMed Links

Gene Details

Gene
WISP1
Gene Name
WNT1 inducible signaling pathway protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204869.1 Intron NP_001191798.1
NM_001204870.1 Intron NP_001191799.1
NM_003882.3 Intron NP_003873.1
NM_080838.2 Intron NP_543028.1

View Full Product Details