Product Details

SNP ID
rs2274969
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:124859095 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAACAGAGCAGCCACTAGCCACACA[T/C]GTAGCTGCTGACATTAAAGGTTTAC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
There are no transcripts associated with this gene.

Gene
RPL35
Gene Name
ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007209.3 Intron NP_009140.1
Gene
WDR38
Gene Name
WD repeat domain 38
There are no transcripts associated with this gene.

View Full Product Details