Product Details

SNP ID
rs2293510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56267641 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGACCTTTTGCACTCGTGACTCCCT[A/G]GCCAGAGAGAGCCTAGCGGGCTGCA
Phenotype
MIM: 118950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
There are no transcripts associated with this gene.

Gene
COQ10A
Gene Name
coenzyme Q10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099337.1 Intron NP_001092807.1
NM_144576.3 Intron NP_653177.3
Gene
CS
Gene Name
citrate synthase
There are no transcripts associated with this gene.

View Full Product Details