Product Details
- SNP ID
-
rs2296662
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35776992 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGTCCTGGGCTCGCTTCCCTCCC[C/G]AAACTGCTCACAGGCAAACCCACCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLPSL1
PubMed Links
Gene Details
- Gene
- CLPSL1
- Gene Name
- colipase like 1
There are no transcripts associated with this gene.
- Gene
- CLPSL2
- Gene Name
- colipase like 2
View Full Product Details