Product Details

SNP ID
rs2293582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:32718211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGAGTGGGCGGAGGGAAGAGG[A/G]CCGCAAACCAACCCAGGACCCGCTC
Phenotype
MIM: 603054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GREM1 PubMed Links

Gene Details

Gene
GREM1
Gene Name
gremlin 1, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191322.1 Intron NP_001178251.1
NM_001191323.1 Intron NP_001178252.1
NM_013372.6 Intron NP_037504.1
XM_005254301.2 Intron XP_005254358.1
XM_017022077.1 Intron XP_016877566.1
Gene
LOC100131315
Gene Name
uncharacterized LOC100131315
There are no transcripts associated with this gene.

View Full Product Details