Product Details

SNP ID
rs2296917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31844669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGGGTTCCTTGTGGTCAGTTG[C/G]CAGGGGCAGTGGGGCCCGGAGTGAG
Phenotype
MIM: 616776 MIM: 602939 MIM: 606566
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 949 Intron NP_001012662.1
NM_001320478.1 949 Intron NP_001307407.1
NM_001320479.1 949 Intron NP_001307408.1
NM_080611.4 949 Intron NP_542178.2
NM_177991.2 949 Intron NP_817130.1
XM_017027654.1 949 Intron XP_016883143.1
XM_017027655.1 949 Intron XP_016883144.1
XM_017027656.1 949 Intron XP_016883145.1
XM_017027657.1 949 Intron XP_016883146.1
XM_017027658.1 949 Intron XP_016883147.1
XM_017027659.1 949 Intron XP_016883148.1
XM_017027660.1 949 Intron XP_016883149.1
XM_017027661.1 949 Intron XP_016883150.1
XM_017027662.1 949 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004118.3 949 Missense Mutation CCA,GCA P292A NP_004109.1
Gene
MYLK2
Gene Name
myosin light chain kinase 2
There are no transcripts associated with this gene.

View Full Product Details