Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001304476.1 | 1394 | UTR 3 | NP_001291405.1 | ||
NM_001304477.1 | 1394 | UTR 3 | NP_001291406.1 | ||
NM_014178.7 | 1394 | UTR 3 | NP_054897.4 | ||
XM_006720121.3 | 1394 | UTR 3 | XP_006720184.1 | ||
XM_011536669.2 | 1394 | UTR 3 | XP_011534971.1 | ||
XM_017021231.1 | 1394 | UTR 3 | XP_016876720.1 | ||
XM_017021232.1 | 1394 | UTR 3 | XP_016876721.1 | ||
XM_017021233.1 | 1394 | UTR 3 | XP_016876722.1 | ||
XM_017021234.1 | 1394 | UTR 3 | XP_016876723.1 | ||
XM_017021235.1 | 1394 | UTR 3 | XP_016876724.1 | ||
XM_017021236.1 | 1394 | UTR 3 | XP_016876725.1 | ||
XM_017021237.1 | 1394 | UTR 3 | XP_016876726.1 | ||
XM_017021238.1 | 1394 | UTR 3 | XP_016876727.1 | ||
XM_017021239.1 | 1394 | UTR 3 | XP_016876728.1 | ||
XM_017021240.1 | 1394 | UTR 3 | XP_016876729.1 | ||
XM_017021241.1 | 1394 | UTR 3 | XP_016876730.1 | ||
XM_017021242.1 | 1394 | UTR 3 | XP_016876731.1 |