Product Details

SNP ID
rs2294012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142702857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCAAACAGCAACCCCTAGACC[C/T]GCATTCCCAGTGCATATGGACAGGC
Phenotype
MIM: 615093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
LY6K
Gene Name
lymphocyte antigen 6 complex, locus K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160354.1 648 Silent Mutation CCC,CCT P77P NP_001153826.1
NM_001160355.1 648 Intron NP_001153827.1
NM_017527.3 648 Intron NP_059997.3
XM_005250989.4 648 Intron XP_005251046.1
XM_005250990.4 648 UTR 3 XP_005251047.1

View Full Product Details