Product Details

SNP ID
rs2297760
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:152514558 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTCAGTGGAGCCAGGGTCTGGTAAG[C/T]CTCCTAGCAGGGTCTGGGGGAGAGG
Phenotype
MIM: 612619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRCT1 PubMed Links

Gene Details

Gene
CRCT1
Gene Name
cysteine rich C-terminal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019060.2 Intron NP_061933.1
XM_011509656.2 Intron XP_011507958.1
Gene
LCE5A
Gene Name
late cornified envelope 5A
There are no transcripts associated with this gene.

View Full Product Details