Product Details

SNP ID
rs2297807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63944500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGGTGACCGGGGGCTGGGCCG[C/T]TGGCCTGCCCGACACCCACCCAACA
Phenotype
MIM: 611203 MIM: 610866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5 PubMed Links

Gene Details

Gene
DNAJC5
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
There are no transcripts associated with this gene.

Gene
MIR1914
Gene Name
microRNA 1914
There are no transcripts associated with this gene.

Gene
MIR647
Gene Name
microRNA 647
There are no transcripts associated with this gene.

Gene
UCKL1
Gene Name
uridine-cytidine kinase 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193379.1 Intron NP_001180308.1
NM_017859.3 Intron NP_060329.2
XM_005260216.2 Intron XP_005260273.1
XM_006723806.2 Intron XP_006723869.1
XM_006723807.2 Intron XP_006723870.1
XM_006723808.1 Intron XP_006723871.1
XM_006723809.1 Intron XP_006723872.1
XM_006723810.1 Intron XP_006723873.1
XM_006723811.3 Intron XP_006723874.1
XM_011528868.2 Intron XP_011527170.1
XM_011528869.1 Intron XP_011527171.1
XM_011528870.1 Intron XP_011527172.1
XM_011528871.1 Intron XP_011527173.1
XM_011528872.1 Intron XP_011527174.1
XM_011528873.2 Intron XP_011527175.1
XM_017027894.1 Intron XP_016883383.1
XM_017027895.1 Intron XP_016883384.1
XM_017027896.1 Intron XP_016883385.1
XM_017027897.1 Intron XP_016883386.1
XM_017027898.1 Intron XP_016883387.1
XM_017027899.1 Intron XP_016883388.1
XM_017027900.1 Intron XP_016883389.1
XM_017027901.1 Intron XP_016883390.1
XM_017027902.1 Intron XP_016883391.1
XM_017027903.1 Intron XP_016883392.1
XM_017027904.1 Intron XP_016883393.1
XM_017027905.1 Intron XP_016883394.1
XM_017027906.1 Intron XP_016883395.1
XM_017027907.1 Intron XP_016883396.1
XM_017027908.1 Intron XP_016883397.1
Gene
UCKL1-AS1
Gene Name
UCKL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details