Product Details

SNP ID
rs2298303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72782098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCCTTCTGGACGCCACCTCACA[A/G]CGTGTGTTGCTTTCCGCTCCTCGGA
Phenotype
MIM: 607533 MIM: 607532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIS3 PubMed Links

Gene Details

Gene
DIS3
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128226.2 35 UTR 5 NP_001121698.1
NM_001322348.1 35 UTR 5 NP_001309277.1
NM_001322349.1 35 UTR 5 NP_001309278.1
NM_014953.4 35 UTR 5 NP_055768.3
Gene
PIBF1
Gene Name
progesterone immunomodulatory binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006346.2 35 UTR 5 NP_006337.2
XM_011534881.2 35 UTR 5 XP_011533183.1
XM_011534882.2 35 UTR 5 XP_011533184.1
XM_011534884.2 35 UTR 5 XP_011533186.1
XM_011534885.2 35 Intron XP_011533187.1
XM_011534886.2 35 UTR 5 XP_011533188.1
XM_017020350.1 35 Intron XP_016875839.1
XM_017020351.1 35 UTR 5 XP_016875840.1
XM_017020352.1 35 Intron XP_016875841.1
XM_017020353.1 35 Intron XP_016875842.1
XM_017020354.1 35 Intron XP_016875843.1

View Full Product Details