Product Details

SNP ID
rs2295227
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:18904523 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CTAAAAATCAGGTAGATGCAAAAAC[A/G]TCCACAAGGGGTCTTGCAGGGAAGG
Phenotype
MIM: 606811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH4A1 PubMed Links

Gene Details

Gene
ALDH4A1
Gene Name
aldehyde dehydrogenase 4 family member A1
There are no transcripts associated with this gene.

Gene
IFFO2
Gene Name
intermediate filament family orphan 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136265.1 5592 UTR 3 NP_001129737.1
XM_011540630.2 5592 UTR 3 XP_011538932.1
Gene
MIR1290
Gene Name
microRNA 1290
There are no transcripts associated with this gene.

View Full Product Details