Product Details

SNP ID
rs2298633
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:23383748 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GTGACCACACTCTGTTCCTTCAGGA[C/G]ACTTGGCAGGAAGGAGCACTCTGGG
Phenotype
MIM: 604128
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCEA3 PubMed Links

Gene Details

Gene
TCEA3
Gene Name
transcription elongation factor A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003196.2 1478 Intron NP_003187.1
XM_006710864.1 1478 UTR 3 XP_006710927.1
XM_011542053.2 1478 Intron XP_011540355.1
XM_017002200.1 1478 Missense Mutation CTC,GTC L463V XP_016857689.1
XM_017002201.1 1478 Missense Mutation CTC,GTC L442V XP_016857690.1
XM_017002202.1 1478 Missense Mutation CTC,GTC L442V XP_016857691.1
XM_017002203.1 1478 Missense Mutation CTC,GTC L426V XP_016857692.1
XM_017002204.1 1478 Intron XP_016857693.1

View Full Product Details