Product Details

SNP ID
rs2298693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46230278 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAAGGCTTAAATCTGCAAGCAAG[A/C]GGGCAAGCTACTAAATCACCCGTTA
Phenotype
MIM: 600909 MIM: 603294
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LSS PubMed Links

Gene Details

Gene
LSS
Gene Name
lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)
There are no transcripts associated with this gene.

Gene
MCM3AP
Gene Name
minichromosome maintenance complex component 3 associated protein
There are no transcripts associated with this gene.

Gene
MCM3AP-AS1
Gene Name
MCM3AP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details