Product Details

SNP ID
rs2295606
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:33862951 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TAGTCTTAGGCAAGTTTCCTTGTCC[A/G]GCTCAGCTGCTGAGACCACATATCT
Phenotype
MIM: 608398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSMD2 PubMed Links

Gene Details

Gene
CSMD2
Gene Name
CUB and Sushi multiple domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281956.1 503 Intron NP_001268885.1
NM_052896.4 503 Intron NP_443128.2
XM_017000185.1 503 Intron XP_016855674.1
XM_017000186.1 503 Intron XP_016855675.1
XM_017000187.1 503 Intron XP_016855676.1
XM_017000188.1 503 Intron XP_016855677.1
XM_017000189.1 503 Intron XP_016855678.1
XM_017000190.1 503 Intron XP_016855679.1
XM_017000191.1 503 Intron XP_016855680.1
XM_017000192.1 503 Intron XP_016855681.1
XM_017000193.1 503 Intron XP_016855682.1
XM_017000194.1 503 Intron XP_016855683.1
Gene
CSMD2-AS1
Gene Name
CSMD2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
HMGB4
Gene Name
high mobility group box 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145205.5 503 UTR 5 NP_660206.2

View Full Product Details