Product Details

SNP ID
rs2298819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77662426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATACAAAAAGTGGTGAGTGTATA[A/C]TCTAAAGTGGTTATATACTAAGCTC
Phenotype
MIM: 608522
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RSF1 PubMed Links

Gene Details

Gene
RSF1
Gene Name
remodeling and spacing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016578.3 8711 Intron NP_057662.3
XM_005274051.2 8711 UTR 3 XP_005274108.1
XM_017017923.1 8711 UTR 3 XP_016873412.1

View Full Product Details