Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286771.2 | 7055 | Missense Mutation | CAT,TAT | H2447Y | NP_001273700.1 |
NM_015574.1 | 7055 | Missense Mutation | CAT,TAT | H2559Y | NP_056389.1 |
NM_032217.4 | 7055 | Missense Mutation | CAT,TAT | H2560Y | NP_115593.3 |
NM_198889.2 | 7055 | Missense Mutation | CAT,TAT | H2309Y | NP_942592.1 |
XM_005265671.3 | 7055 | Missense Mutation | CAT,TAT | H2308Y | XP_005265728.1 |
XM_005265672.3 | 7055 | Intron | XP_005265729.1 | ||
XM_005265673.3 | 7055 | Intron | XP_005265730.1 | ||
XM_017008011.1 | 7055 | Missense Mutation | CAT,TAT | H2446Y | XP_016863500.1 |
XM_017008012.1 | 7055 | Missense Mutation | CAT,TAT | H2196Y | XP_016863501.1 |
XM_017008013.1 | 7055 | Missense Mutation | CAT,TAT | H2195Y | XP_016863502.1 |
XM_017008014.1 | 7055 | Intron | XP_016863503.1 | ||
XM_017008015.1 | 7055 | Intron | XP_016863504.1 | ||
XM_017008016.1 | 7055 | Intron | XP_016863505.1 | ||
XM_017008017.1 | 7055 | Intron | XP_016863506.1 |