Product Details

SNP ID
rs2306111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138145233 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTTCTGACACAGGCTTCAGAAA[C/T]GGACTGCTGAACCTGTTAAGGGACA
Phenotype
MIM: 602848 MIM: 603575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRD8 PubMed Links

Gene Details

Gene
BRD8
Gene Name
bromodomain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164326.1 3484 Intron NP_001157798.1
NM_001300961.1 3484 Intron NP_001287890.1
NM_001300962.1 3484 Intron NP_001287891.1
NM_001300966.1 3484 Intron NP_001287895.1
NM_006696.3 3484 Intron NP_006687.3
NM_139199.1 3484 Silent Mutation CCA,CCG P1127P NP_631938.1
XM_005271855.2 3484 Silent Mutation CCA,CCG P1200P XP_005271912.1
XM_005271856.2 3484 Silent Mutation CCA,CCG P1185P XP_005271913.1
XM_005271857.2 3484 Silent Mutation CCA,CCG P1130P XP_005271914.1
XM_005271859.3 3484 Silent Mutation CCA,CCG P1057P XP_005271916.1
XM_005271860.3 3484 Intron XP_005271917.1
XM_005271861.3 3484 Intron XP_005271918.1
XM_011543109.1 3484 Silent Mutation CCA,CCG P1146P XP_011541411.1
XM_011543110.1 3484 Silent Mutation CCA,CCG P1067P XP_011541412.1
XM_017008969.1 3484 Silent Mutation CCA,CCG P1073P XP_016864458.1
XM_017008970.1 3484 Intron XP_016864459.1
XM_017008971.1 3484 Intron XP_016864460.1
XM_017008972.1 3484 Intron XP_016864461.1
XM_017008973.1 3484 Intron XP_016864462.1
XM_017008974.1 3484 Intron XP_016864463.1
XM_017008975.1 3484 Intron XP_016864464.1
XM_017008976.1 3484 Intron XP_016864465.1
Gene
NME5
Gene Name
NME/NM23 family member 5
There are no transcripts associated with this gene.

View Full Product Details