Product Details
- SNP ID
-
rs2306211
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:121432937 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATACCTGAACAACATCTTCTTCT[A/G]CCACTTCATATAGGAGTTCATCATG
- Phenotype
-
MIM: 604419
MIM: 609381
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POLQ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3218634] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POLQ
- Gene Name
- DNA polymerase theta
- Gene
- STXBP5L
- Gene Name
- syntaxin binding protein 5 like
There are no transcripts associated with this gene.
View Full Product Details