Product Details

SNP ID
rs3003572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128209149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTCAGAGCACGCAGCTGCCAC[A/G]TGCTAAGCACACACTGCCTGTGGGC
Phenotype
MIM: 611420 MIM: 602377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIZ1 PubMed Links

Gene Details

Gene
CIZ1
Gene Name
CDKN1A interacting zinc finger protein 1
There are no transcripts associated with this gene.

Gene
DNM1
Gene Name
dynamin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005336.2 Intron NP_001005336.1
NM_001288737.1 Intron NP_001275666.1
NM_001288738.1 Intron NP_001275667.1
NM_001288739.1 Intron NP_001275668.1
NM_004408.3 Intron NP_004399.2
XM_005251763.2 Intron XP_005251820.1
XM_005251768.2 Intron XP_005251825.1
XM_005251769.2 Intron XP_005251826.1
XM_006716992.2 Intron XP_006717055.1
XM_006716993.1 Intron XP_006717056.1
XM_011518335.2 Intron XP_011516637.1
XM_011518336.2 Intron XP_011516638.1
XM_017014369.1 Intron XP_016869858.1
XM_017014370.1 Intron XP_016869859.1
XM_017014371.1 Intron XP_016869860.1
XM_017014372.1 Intron XP_016869861.1
XM_017014373.1 Intron XP_016869862.1

View Full Product Details