Product Details

SNP ID
rs2306828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45136405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGATGCCCTGGGTGGGACCCATC[A/C]CCACCTCTGCCCTTTCCAAGCAGGG
Phenotype
MIM: 608795
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACBD4 PubMed Links

Gene Details

Gene
ACBD4
Gene Name
acyl-CoA binding domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135705.2 Intron NP_001129177.1
NM_001135706.2 Intron NP_001129178.1
NM_001135707.2 Intron NP_001129179.1
NM_001321352.1 Intron NP_001308281.1
NM_001321353.1 Intron NP_001308282.1
NM_024722.3 Intron NP_078998.1
XM_006722085.2 Intron XP_006722148.1
XM_011525259.2 Intron XP_011523561.1
XM_011525261.2 Intron XP_011523563.1
XM_017025084.1 Intron XP_016880573.1
XM_017025085.1 Intron XP_016880574.1
XM_017025086.1 Intron XP_016880575.1
XM_017025087.1 Intron XP_016880576.1
XM_017025088.1 Intron XP_016880577.1
XM_017025089.1 Intron XP_016880578.1
XM_017025090.1 Intron XP_016880579.1
XM_017025091.1 Intron XP_016880580.1
XM_017025092.1 Intron XP_016880581.1
XM_017025093.1 Intron XP_016880582.1
XM_017025094.1 Intron XP_016880583.1
XM_017025095.1 Intron XP_016880584.1
XM_017025096.1 Intron XP_016880585.1
XM_017025097.1 Intron XP_016880586.1
XM_017025098.1 Intron XP_016880587.1
XM_017025099.1 Intron XP_016880588.1
XM_017025100.1 Intron XP_016880589.1
XM_017025101.1 Intron XP_016880590.1
Gene
PLCD3
Gene Name
phospholipase C delta 3
There are no transcripts associated with this gene.

View Full Product Details