Product Details

SNP ID
rs2306897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6450903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCCCCCTATACCTGCAAAGTGG[A/C]GGTCAGTGGCCCCACGGCCCCCAAA
Phenotype
MIM: 142290 MIM: 605493
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HPX PubMed Links

Gene Details

Gene
HPX
Gene Name
hemopexin
There are no transcripts associated with this gene.

Gene
TRIM3
Gene Name
tripartite motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248006.1 2035 Missense Mutation NP_001234935.1
NM_001248007.1 2035 Missense Mutation NP_001234936.1
NM_006458.3 2035 Missense Mutation NP_006449.2
NM_033278.3 2035 Missense Mutation NP_150594.2
XM_011519844.1 2035 Missense Mutation XP_011518146.1
XM_011519845.2 2035 Missense Mutation XP_011518147.1
XM_017017098.1 2035 Missense Mutation XP_016872587.1
XM_017017099.1 2035 Missense Mutation XP_016872588.1

View Full Product Details