Product Details

SNP ID
rs3006013
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:244471635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTCATAGAAACAAAACCATACT[A/G]TATATAGTCTTTTGGGGTCAGTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf101 PubMed Links

Gene Details

Gene
C1orf101
Gene Name
chromosome 1 open reading frame 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130957.1 Intron NP_001124429.1
NM_001242340.1 Intron NP_001229269.1
NM_173807.4 Intron NP_776168.1
XM_011544138.2 Intron XP_011542440.1
XM_011544139.2 Intron XP_011542441.1
XM_011544142.2 Intron XP_011542444.1
XM_011544144.2 Intron XP_011542446.1
XM_011544145.2 Intron XP_011542447.1
XM_011544149.2 Intron XP_011542451.1
XM_011544151.2 Intron XP_011542453.1
XM_017000943.1 Intron XP_016856432.1
XM_017000944.1 Intron XP_016856433.1
XM_017000945.1 Intron XP_016856434.1
XM_017000946.1 Intron XP_016856435.1
XM_017000947.1 Intron XP_016856436.1
XM_017000948.1 Intron XP_016856437.1
XM_017000949.1 Intron XP_016856438.1
XM_017000950.1 Intron XP_016856439.1
XM_017000951.1 Intron XP_016856440.1
XM_017000952.1 Intron XP_016856441.1
XM_017000953.1 Intron XP_016856442.1

View Full Product Details