Product Details
- SNP ID
-
hCV16200554
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:131745567 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCCTTTGGGCAATGTGCTCTCTG[C/T]CCTGCTGGTGGTGTGCCTTCCCTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf27A
PubMed Links
Gene Details
- Gene
- C2orf27A
- Gene Name
- chromosome 2 open reading frame 27A
There are no transcripts associated with this gene.
View Full Product Details