Product Details

SNP ID
rs2335156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2696990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGGGATTGGCTCAGAAACGGGC[A/G]CTAGACACAGACCAAGGCCCTGCCG
Phenotype
MIM: 611285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD5 PubMed Links

Gene Details

Gene
KCTD5
Gene Name
potassium channel tetramerization domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018992.3 Intron NP_061865.1

View Full Product Details