Product Details

SNP ID
rs2341537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55422975 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGCTATTTATGCAAAAATTATT[A/T]AAAAAAAAGCCAACAGGAAAAAATA
Phenotype
MIM: 608964
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TBPL2 PubMed Links

Gene Details

Gene
TBPL2
Gene Name
TATA-box binding protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199047.2 Intron NP_950248.1

View Full Product Details