Product Details

SNP ID
rs2370971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54581082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAACATAGGGGTGGGAGGAAGA[A/G]GTGGCATTCATAAAGGTGTTGGGGA
Phenotype
MIM: 171891 MIM: 613246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE1B PubMed Links

Gene Details

Gene
PDE1B
Gene Name
phosphodiesterase 1B
There are no transcripts associated with this gene.

Gene
PPP1R1A
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006741.3 Intron NP_006732.3
XM_005268995.4 Intron XP_005269052.1
XM_006719471.3 Intron XP_006719534.1

View Full Product Details