Product Details
- SNP ID
-
rs2367370
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:86788961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCCAAGGACGAACCTCAAACTG[A/G]TGGTGGTTTTGACTAACCCTCCTGG
- Phenotype
-
MIM: 186910
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD8A
PubMed Links
Gene Details
- Gene
- CD8A
- Gene Name
- CD8a molecule
View Full Product Details