Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012446.3 | 67 | Intron | NP_001012448.2 | ||
XM_006716768.3 | 67 | Intron | XP_006716831.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042589.2 | 67 | Intron | NP_001036054.1 | ||
NM_001042590.2 | 67 | Intron | NP_001036055.1 | ||
NM_016446.3 | 67 | Intron | NP_057530.2 | ||
XM_005251483.4 | 67 | Intron | XP_005251540.3 | ||
XM_011517901.2 | 67 | Intron | XP_011516203.1 | ||
XM_011517902.2 | 67 | Intron | XP_011516204.1 | ||
XM_011517903.2 | 67 | Intron | XP_011516205.1 | ||
XM_011517904.2 | 67 | UTR 5 | XP_011516206.1 | ||
XM_011517905.1 | 67 | Intron | XP_011516207.1 | ||
XM_011517908.2 | 67 | Intron | XP_011516210.1 | ||
XM_011517910.2 | 67 | Intron | XP_011516212.1 | ||
XM_011517911.2 | 67 | Intron | XP_011516213.1 | ||
XM_011517912.2 | 67 | Intron | XP_011516214.1 | ||
XM_011517913.2 | 67 | Intron | XP_011516215.1 | ||
XM_011517914.2 | 67 | Intron | XP_011516216.1 | ||
XM_011517915.2 | 67 | Intron | XP_011516217.1 | ||
XM_011517916.2 | 67 | Intron | XP_011516218.1 | ||
XM_011517918.2 | 67 | Intron | XP_011516220.1 | ||
XM_017014805.1 | 67 | Intron | XP_016870294.1 | ||
XM_017014806.1 | 67 | Intron | XP_016870295.1 | ||
XM_017014807.1 | 67 | Intron | XP_016870296.1 |