Product Details
- SNP ID
-
rs2446205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5812902 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAAAACAGAGGCACAGGCGCGC[A/G]AACACATGTATGCCGCGGCGTTCAC
- Phenotype
-
MIM: 602018
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NRTN
PubMed Links
Gene Details
- Gene
- NRTN
- Gene Name
- neurturin
View Full Product Details