Product Details
- SNP ID
-
rs2473930
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:34152290 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGGGGGAGGAGGAGGAAACAGCCC[C/T]GTGAACCTGGCTCCAAGAGGGCCTA
- Phenotype
-
MIM: 609221
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABTB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113963484] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABTB2
- Gene Name
- ankyrin repeat and BTB domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145804.2 |
3600 |
UTR 3 |
|
|
NP_665803.2 |
- Gene
- NAT10
- Gene Name
- N-acetyltransferase 10
There are no transcripts associated with this gene.
View Full Product Details