Product Details

SNP ID
rs2459215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124401087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGAAAAACATGACTAGATAAAA[G/T]ACCAGAGGAACTTAACTTTCATTGC
Phenotype
MIM: 613349
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OAT PubMed Links

Gene Details

Gene
OAT
Gene Name
ornithine aminotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000274.3 Intron NP_000265.1
NM_001171814.1 Intron NP_001165285.1
NM_001322965.1 Intron NP_001309894.1
NM_001322966.1 Intron NP_001309895.1
NM_001322967.1 Intron NP_001309896.1
NM_001322968.1 Intron NP_001309897.1
NM_001322969.1 Intron NP_001309898.1
NM_001322970.1 Intron NP_001309899.1
NM_001322971.1 Intron NP_001309900.1
NM_001322974.1 Intron NP_001309903.1
XM_017016279.1 Intron XP_016871768.1

View Full Product Details