Product Details

SNP ID
rs2440199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96144183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGAGAAACGGGTATGCATCTC[C/T]CCTCCCCTCCCCTTCTATCAAAGCC
Phenotype
MIM: 601147
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDF6 PubMed Links

Gene Details

Gene
GDF6
Gene Name
growth differentiation factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001557.2 1848 UTR 3 NP_001001557.1

View Full Product Details