Product Details

SNP ID
rs2517659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30152625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTTGAGTTTGCACCCTGGATG[C/G]CTCCCCTCCTCCCAGGAGACCCAGT
Phenotype
MIM: 605701 MIM: 616976
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM10 PubMed Links

Gene Details

Gene
TRIM10
Gene Name
tripartite motif containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006778.3 3114 UTR 3 NP_006769.2
NM_052828.2 3114 UTR 3 NP_439893.2
XM_011514221.1 3114 UTR 3 XP_011512523.1
XM_011514222.2 3114 UTR 3 XP_011512524.1
XM_011514223.2 3114 UTR 3 XP_011512525.1
XM_011514225.1 3114 UTR 3 XP_011512527.1
Gene
TRIM40
Gene Name
tripartite motif containing 40
There are no transcripts associated with this gene.

View Full Product Details