Product Details

SNP ID
rs2518723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21995883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGCCTGCGGTCTGCCTGGATC[C/T]GTCCTAAACCTCGCGGGCTGGACCC
Phenotype
MIM: 600160 MIM: 600431 MIM: 613149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN2A PubMed Links

Gene Details

Gene
CDKN2A
Gene Name
cyclin dependent kinase inhibitor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000077.4 Intron NP_000068.1
NM_001195132.1 Intron NP_001182061.1
NM_058195.3 Intron NP_478102.2
NM_058197.4 Intron NP_478104.2
XM_005251343.1 Intron XP_005251400.1
XM_011517675.2 Intron XP_011515977.1
XM_011517676.2 Intron XP_011515978.1
XM_011517679.1 Intron XP_011515981.1
Gene
CDKN2B
Gene Name
cyclin dependent kinase inhibitor 2B
There are no transcripts associated with this gene.

Gene
CDKN2B-AS1
Gene Name
CDKN2B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details