Product Details

SNP ID
hCV16247112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:85382167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTCTGTCATTCATAGTGGAGC[A/C]AAATGGCCAGAGTGTCATTTATTGT
Phenotype
MIM: 604686
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKAP13 PubMed Links

Gene Details

Gene
AKAP13
Gene Name
A-kinase anchoring protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270546.1 Intron NP_001257475.1
NM_006738.5 Intron NP_006729.4
NM_007200.4 Intron NP_009131.2
Gene
MIR7706
Gene Name
microRNA 7706
There are no transcripts associated with this gene.

View Full Product Details