Product Details

SNP ID
rs2257791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116884159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACTTAAAATTGTGTAAGCAGGAC[A/G]TATGTAAGTTTTGTGTGTGCAATAG
Phenotype
MIM: 611171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO4 PubMed Links

Gene Details

Gene
ENO4
Gene Name
enolase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242699.1 4265 Intron NP_001229628.1
XM_005269815.3 4265 Intron XP_005269872.1
XM_006717835.3 4265 Intron XP_006717898.1
XM_006717836.3 4265 Intron XP_006717899.1
XM_011539797.2 4265 Intron XP_011538099.1
XM_011539799.2 4265 Intron XP_011538101.1
XM_017016241.1 4265 Intron XP_016871730.1
Gene
SHTN1
Gene Name
shootin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127211.2 4265 UTR 3 NP_001120683.1
NM_001258298.1 4265 UTR 3 NP_001245227.1
NM_001258299.1 4265 Intron NP_001245228.1
NM_001258300.1 4265 Intron NP_001245229.1
NM_018330.6 4265 UTR 3 NP_060800.2
XM_005270002.3 4265 Intron XP_005270059.1
XM_006717931.3 4265 Intron XP_006717994.1
XM_017016462.1 4265 Intron XP_016871951.1

View Full Product Details