Product Details

SNP ID
rs2150468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:38353751 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTACTAATTTTGAACATGATTA[G/T]TAAGAGTACACAGAAGGTTCATCAA
Phenotype
MIM: 610553
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UFM1 PubMed Links

Gene Details

Gene
UFM1
Gene Name
ubiquitin fold modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286703.1 Intron NP_001273632.1
NM_001286704.1 Intron NP_001273633.1
NM_001286705.1 Intron NP_001273634.1
NM_001286706.1 Intron NP_001273635.1
NM_016617.3 Intron NP_057701.1

View Full Product Details