Product Details

SNP ID
rs2528641
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:44215010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGGGCACGCTTATGCCCAACT[A/G]TGATTACCCACTCAATCCTCGTGGC
Phenotype
MIM: 605077 MIM: 609917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMAP1 PubMed Links

Gene Details

Gene
DMAP1
Gene Name
DNA methyltransferase 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034023.1 Intron NP_001029195.1
NM_001034024.1 Intron NP_001029196.1
NM_019100.4 Intron NP_061973.1
XM_005271037.4 Intron XP_005271094.1
XM_005271039.1 Intron XP_005271096.1
XM_005271040.1 Intron XP_005271097.1
XM_006710771.3 Intron XP_006710834.1
XM_011541784.2 Intron XP_011540086.1
XM_011541785.2 Intron XP_011540087.1
XM_017001807.1 Intron XP_016857296.1
XM_017001808.1 Intron XP_016857297.1
Gene
ERI3
Gene Name
ERI1 exoribonuclease family member 3
There are no transcripts associated with this gene.

Gene
ERI3-IT1
Gene Name
ERI3 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details