Product Details
- SNP ID
-
rs2575695
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:103373488 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACATTTATTGAGAGTACTAAGCAT[A/G]TGTTGGGTTAGCATTTTAAAGTAGA
- Phenotype
-
MIM: 610635
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTHRC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77776607] are located under a probe and SNP(s) [rs33982219] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTHRC1
- Gene Name
- collagen triple helix repeat containing 1
View Full Product Details