Product Details
- SNP ID
-
rs2575705
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:96496103 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAAAAACTGCACATGGTTAAGGAG[A/G]CCCTTTTTTTTTGCTGAGTCTTATT
- Phenotype
-
MIM: 142460
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDC2
PubMed Links
Gene Details
- Gene
- SDC2
- Gene Name
- syndecan 2
View Full Product Details