Product Details
- SNP ID
-
rs2576092
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:45116735 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACGCCTCGGCCACGCTATGGTAAG[C/T]GCTGGAGGGAAGGCTGTGTGCACGT
- Phenotype
-
MIM: 606759
MIM: 612771
MIM: 612772
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DUOX2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs560539864] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DUOX2
- Gene Name
- dual oxidase 2
There are no transcripts associated with this gene.
- Gene
- DUOXA1
- Gene Name
- dual oxidase maturation factor 1
- Gene
- DUOXA2
- Gene Name
- dual oxidase maturation factor 2
View Full Product Details