Product Details
- SNP ID
-
rs2506014
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:43081119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCAGCCCCCCATGCTCCCTGCT[C/G]CCCTCCTCCCCTCCTGCAGAGGCAC
- Phenotype
-
MIM: 164761
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RET
PubMed Links
Gene Details
- Gene
- RET
- Gene Name
- ret proto-oncogene
There are no transcripts associated with this gene.
View Full Product Details