Product Details

SNP ID
rs2601779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3959528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTGCAGAAAATTTAACAACCTA[G/T]GTAAATTAAAAAACCTAAGTAAAGG
Phenotype
MIM: 603302
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADCY9 PubMed Links

Gene Details

Gene
ADCY9
Gene Name
adenylate cyclase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001116.3 Intron NP_001107.2
XM_005255079.3 Intron XP_005255136.1
XM_011522353.2 Intron XP_011520655.1
Gene
LOC102724927
Gene Name
uncharacterized LOC102724927
There are no transcripts associated with this gene.

View Full Product Details