Product Details

SNP ID
rs2560877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52269760 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTCCACCCCGAGTAAATTCATG[C/T]GTCCCGTCAGAGTGTTAAAATCGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF766 PubMed Links

Gene Details

Gene
ZNF766
Gene Name
zinc finger protein 766
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010851.2 Intron NP_001010851.1
XM_011527460.1 Intron XP_011525762.1
XM_017027440.1 Intron XP_016882929.1
XM_017027441.1 Intron XP_016882930.1

View Full Product Details